Assay Details
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034732.1 | NP_001029904.1 | 2 - 3 | 215 | 135 | |
GenBank mRNA | BC102971.1 | - | 2 - 3 | 215 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
