Assay Details
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034733.2 | NP_001029905.1 | 2 - 3 | 370 | 145 | |
GenBank mRNA | BC103462.1 | - | 2 - 3 | 371 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
