Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034752.1 | NP_001029924.1 | 8 - 9 | 1071 | 79 | |
GenBank mRNA | BC104616.1 | - | 9 - 10 | 1085 | 79 | |
BT021050.1 | - | 8 - 9 | 1071 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
