Assay Details
Gene Symbol: CENPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034782.1 | NP_001029954.1 | 1 - 2 | 382 | 75 | |
GenBank mRNA | BC126617.1 | - | 1 - 2 | 436 | 75 | |
BT020751.1 | - | 1 - 2 | 382 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126617.1, NM_001034782.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
