Assay Details
Gene Symbol: CENPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034782.1 | NP_001029954.1 | 4 - 5 | 1364 | 82 | |
GenBank mRNA | BC126617.1 | - | 4 - 5 | 1418 | 82 | |
BT020751.1 | - | 4 - 5 | 1364 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
