Assay Details
Gene Symbol: CRIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034803.2 | NP_001029975.1 | 1 - 2 | 94 | 91 | |
GenBank mRNA | BC102887.1 | - | 1 - 2 | 95 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102887.1, NM_001034803.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
