Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035075.1 | NP_001030247.1 | 2 - 3 | 220 | 65 | |
GenBank mRNA | BC122801.1 | - | 1 - 2 | 65 | ||
BT021626.1 | - | 2 - 3 | 220 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122801.1, NM_001035075.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
