Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035075.1 | NP_001030247.1 | 5 - 6 | 605 | 101 | |
GenBank mRNA | BC122801.1 | - | 4 - 5 | 435 | 101 | |
BT021626.1 | - | 5 - 6 | 605 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
