Assay Details
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035287.1 | NP_001030364.1 | 6 - 7 | 776 | 114 | |
GenBank mRNA | BC118173.1 | - | 5 - 6 | 1686 | 114 | |
BT021159.1 | - | 6 - 7 | 776 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
