Assay Details
Gene Symbol: TEX261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035300.1 | NP_001030377.1 | 2 - 3 | 698 | 108 | |
GenBank mRNA | BC133312.1 | - | 3 - 4 | 384 | 108 | |
BT021693.1 | - | 2 - 3 | 698 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
