Assay Details
Gene Symbol: ADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035468.2 | NP_001030545.1 | 1 - 2 | 194 | 103 | |
GenBank mRNA | BC103235.1 | - | 1 - 2 | 195 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
