Assay Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075790.1 | NP_001069258.1 | 11 - 12 | 1549 | 153 | |
GenBank mRNA | BT021737.1 | - | 11 - 12 | 1549 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
