Assay Details
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077040.1 | NP_001070508.1 | 1 - 2 | 124 | 64 | |
GenBank mRNA | BT021572.1 | - | 1 - 2 | 124 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001077040.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
