Assay Details
Gene Symbol: WBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082441.1 | NP_001075910.1 | 6 - 7 | 1068 | 111 | |
GenBank mRNA | AF322215.1 | - | 6 - 7 | 1068 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
