Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083445.1 | NP_001076914.1 | 3 - 4 | 587 | 71 | |
GenBank mRNA | BC133573.1 | - | 3 - 4 | 606 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
