Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083445.1 | NP_001076914.1 | 6 - 7 | 1026 | 141 | |
GenBank mRNA | BC133573.1 | - | 6 - 7 | 1045 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
