Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083445.1 | NP_001076914.1 | 7 - 8 | 1128 | 100 | |
GenBank mRNA | BC133573.1 | - | 7 - 8 | 1147 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
