Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174089.1 | NP_776514.1 | 3 - 4 | 364 | 66 | |
GenBank mRNA | AF072807.1 | - | 3 - 4 | 294 | 66 | |
EU276077.1 | - | 3 - 4 | 337 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
