Assay Details
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174117.1 | NP_776542.1 | 21 - 22 | 2757 | 143 | |
GenBank mRNA | AB059399.2 | - | 21 - 22 | 2757 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
