Assay Details
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174117.1 | NP_776542.1 | 24 - 25 | 3309 | 97 | |
GenBank mRNA | AB059399.2 | - | 24 - 25 | 3309 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
