Assay Details
Gene Symbol: AKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174236.1 | NP_776661.1 | 1 | 853 | 73 | |
GenBank mRNA | M60292.1 | - | 1 | 343 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
