Assay Details
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174251.1 | NP_776676.1 | 1 - 2 | 132 | 80 | |
GenBank mRNA | BC102269.1 | - | 1 - 2 | 140 | 80 | |
L35045.1 | - | 1 - 2 | 132 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
