Assay Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174315.3 | NP_776740.1 | 2 - 3 | 295 | 65 | |
GenBank mRNA | AF059507.1 | - | 2 - 3 | 303 | 65 | |
BC105196.1 | - | 2 - 3 | 313 | 65 | ||
BT020981.1 | - | 2 - 3 | 295 | 65 | ||
U55188.1 | - | 2 - 3 | 315 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
