Assay Details
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174334.3 | NP_776759.1 | 1 - 2 | 41 | 65 | |
GenBank mRNA | AB002156.1 | - | 1 - 2 | 42 | 65 | |
BC110264.1 | - | 1 - 2 | 39 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110264.1, NM_174334.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
