Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174340.2 | NP_776765.1 | 1 - 2 | 138 | 137 | |
GenBank mRNA | AF366940.1 | - | 1 - 2 | 138 | 137 | |
BC114199.1 | - | 1 - 2 | 109 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114199.1, NM_174340.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
