Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174340.2 | NP_776765.1 | 4 - 5 | 458 | 83 | |
GenBank mRNA | AF366940.1 | - | 4 - 5 | 458 | 83 | |
BC114199.1 | - | 4 - 5 | 429 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
