Assay Details
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174347.2 | NP_776772.1 | 1 - 2 | 114 | 80 | |
GenBank mRNA | AF411973.1 | - | 1 - 2 | 114 | 80 | |
BT021754.1 | - | 1 - 2 | 78 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174347.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
