Assay Details
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174347.2 | NP_776772.1 | 2 - 3 | 1095 | 68 | |
GenBank mRNA | AF411973.1 | - | 2 - 3 | 1095 | 68 | |
BC102042.1 | - | 2 - 3 | 1090 | 68 | ||
BT021754.1 | - | 2 - 3 | 1059 | 68 | ||
BT021810.1 | - | 2 - 3 | 1068 | 68 | ||
BT026155.1 | - | 2 - 3 | 1062 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
