Assay Details
Gene Symbol: LOXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174383.2 | NP_776808.1 | 3 - 4 | 1532 | 69 | |
GenBank mRNA | AF421185.1 | - | 3 - 4 | 1532 | 69 | |
BC126600.1 | - | 3 - 4 | 1716 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
