Assay Details
Gene Symbol: MYOZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174397.2 | NP_776822.1 | 1 - 2 | 59 | 83 | |
GenBank mRNA | AF359567.1 | - | 1 - 2 | 59 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174397.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
