Assay Details
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174471.3 | NP_776896.1 | 5 - 6 | 540 | 75 | |
GenBank mRNA | AF144763.1 | - | 4 - 5 | 388 | 75 | |
AY295346.1 | - | 5 - 6 | 540 | 75 | ||
BC102489.1 | - | 5 - 6 | 506 | 75 | ||
KX834564.1 | - | 4 - 5 | 458 | 75 | ||
M60073.1 | - | 4 - 5 | 464 | 75 | ||
S70841.1 | - | 4 - 5 | 460 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
