Assay Details
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174485.4 | NP_776910.2 | 1 - 2 | 73 | 115 | |
GenBank mRNA | BC102113.1 | - | 1 - 2 | 75 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102113.1, NM_174485.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
