Assay Details
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174546.2 | NP_776971.1 | 3 - 4 | 651 | 71 | |
GenBank mRNA | X95352.1 | - | 678 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
