Assay Details
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174580.3 | NP_777005.2 | 4 - 5 | 818 | 81 | |
GenBank mRNA | DQ126156.1 | - | 1 - 2 | 96 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
