Assay Details
Gene Symbol: DHRS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174733.2 | NP_777158.1 | 1 - 2 | 153 | 119 | |
GenBank mRNA | AF362071.1 | - | 1 - 2 | 153 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174733.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
