Assay Details
Gene Symbol: DHRS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174733.2 | NP_777158.1 | 2 - 3 | 523 | 67 | |
GenBank mRNA | AF362071.1 | - | 2 - 3 | 523 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
