Assay Details
Gene Symbol: GGTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177511.2 | NP_803477.1 | 3 - 4 | 459 | 131 | |
GenBank mRNA | BC151691.1 | - | 2 - 3 | 171 | 131 | |
J04989.1 | - | 3 - 4 | 459 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_177511.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
