Assay Details
Gene Symbol: SOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177523.2 | NP_803489.1 | 1 - 2 | 38 | 109 | |
GenBank mRNA | AY183452.1 | - | 1 - 2 | 38 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_177523.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
