Assay Details
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178320.2 | NP_847890.1 | 4 - 5 | 378 | 106 | |
GenBank mRNA | AB098778.1 | - | 1 - 2 | 173 | 105 | |
AB099010.1 | - | 4 - 5 | 378 | 105 | ||
AY247029.1 | - | 4 - 5 | 378 | 106 | ||
AY911355.1 | - | 4 - 5 | 413 | 105 | ||
BC105173.1 | - | 415 | 105 | |||
MK309342.1 | - | 1 - 2 | 145 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
