Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178572.2 | NP_848667.1 | 5 - 6 | 559 | 69 | |
GenBank mRNA | AY240020.1 | - | 5 - 6 | 559 | 69 | |
BC103260.1 | - | 5 - 6 | 613 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
