Assay Details
Gene Symbol: TAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181017.2 | NP_851360.1 | 2 - 3 | 181 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
