Assay Details
Gene Symbol: RHEBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181668.1 | NP_858054.1 | 1 - 2 | 145 | 74 | |
GenBank mRNA | AY293823.1 | - | 1 - 2 | 145 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
