Assay Details
Gene Symbol: MRPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201606.2 | NP_963900.1 | 3 - 4 | 377 | 81 | |
GenBank mRNA | AB116379.1 | - | 3 - 4 | 392 | 81 | |
BC134767.1 | - | 3 - 4 | 401 | 81 | ||
BC142005.1 | - | 3 - 4 | 401 | 81 | ||
BT020735.1 | - | 3 - 4 | 354 | 81 | ||
BT020755.1 | - | 3 - 4 | 389 | 81 | ||
BT020982.1 | - | 3 - 4 | 377 | 81 | ||
BT030597.1 | - | 3 - 4 | 448 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
