Assay Details
Gene Symbol: MYD88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014382.2 | NP_001014404.1 | 3 - 4 | 681 | 60 | |
GenBank mRNA | AJ853453.1 | - | 3 - 4 | 648 | 60 | |
AY634627.1 | - | 1 - 2 | 31 | 60 | ||
AY934808.2 | - | 2 - 3 | 523 | 60 | ||
BC102851.1 | - | 3 - 4 | 681 | 60 | ||
KT601039.1 | - | 3 - 4 | 644 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
