Assay Details
Gene Symbol: HNRNPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014860.3 | NP_001014860.1 | 2 - 3 | 115 | 84 | |
GenBank mRNA | BC102955.1 | - | 2 - 3 | 117 | 84 | |
BT020929.1 | - | 2 - 3 | 96 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102955.1, NM_001014860.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
