Assay Details
Gene Symbol: RPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015556.2 | NP_001015556.1 | 1 | 111 | 62 | |
GenBank mRNA | BC104507.1 | - | 1 | 111 | 62 | |
BT021047.1 | - | 2 - 3 | 110 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
