Assay Details
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034214.2 | NP_001029386.1 | 1 | 417 | 91 | |
GenBank mRNA | BC103189.1 | - | 1 | 418 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
