Assay Details
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034270.1 | NP_001029442.1 | 3 - 4 | 392 | 64 | |
GenBank mRNA | BC102280.1 | - | 3 - 4 | 392 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
