Assay Details
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034299.1 | NP_001029471.1 | 2 - 3 | 229 | 112 | |
GenBank mRNA | AB098924.1 | - | 2 - 3 | 181 | 112 | |
BC102308.1 | - | 2 - 3 | 229 | 112 | ||
BT021601.1 | - | 2 - 3 | 159 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
