Assay Details
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034311.2 | NP_001029483.1 | 1 - 2 | 67 | 85 | |
GenBank mRNA | BC103413.1 | - | 1 - 2 | 68 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103413.1, NM_001034311.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
